Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,6-diiodohexane 97%, Acros Organics

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
629-09-4
C6H12I2
337.971
MFCD00001103
QLIMAARBRDAYGQ-UHFFFAOYSA-N
hexamethylene diiodide,hexane, 1,6-diiodo,1,6-diiodohexane, stabilized with copper,1,6-di-iodohexane,1,6-diiodo hexane,pubchem3846,hexamethyleneiodide,1,6-bis iodanyl hexane,acmc-209tt7,ksc354g6d
12373
1,6-diiodohexane
C(CCCI)CCI
629-09-4
C6H12I2
337.971
MFCD00001103
QLIMAARBRDAYGQ-UHFFFAOYSA-N
hexamethylene diiodide,hexane, 1,6-diiodo,1,6-diiodohexane, stabilized with copper,1,6-di-iodohexane,1,6-diiodo hexane,pubchem3846,hexamethyleneiodide,1,6-bis iodanyl hexane,acmc-209tt7,ksc354g6d
12373
1,6-diiodohexane
C(CCCI)CCI
Packaging Glass bottle
Stabilizer copper
Refractive Index 1.5850 to 1.5870 (20°C, 589nm)
Quantity 25g
Color Yellow
Infrared Spectrum Authentic
Density 2.0520g/mL
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula I(CH2)6I
Formula Weight 337.96
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material 1, 6-Diiodohexane
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.