Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,6-Diiodoperfluorohexane, 97%

Click to view available options
Quantity:
5g
25g
375-80-4
C6F12I2
553.856
MFCD00042264
JOQDDLBOAIKFQX-UHFFFAOYSA-N
1,6-diiodoperfluorohexane,1,6-diiodododecafluorohexane,dodecafluoro-1,6-diiodohexane,1,6-diiododecafluorohexane,perfluoro-1,6-diiodohexane,hexane, 1,1,2,2,3,3,4,4,5,5,6,6-dodecafluoro-1,6-diiodo,acmc-209iu0,1,1,2,2,3,3,4,4,5,5,6,6-dodecafluoro-1,6-diiodo-hexane,hexane,1,1,2,2,3,3,4,4,5,5,6,6-dodecafluoro-1,6-diiodo
78994
1,1,2,2,3,3,4,4,5,5,6,6-dodecafluoro-1,6-diiodohexane
C(C(C(C(F)(F)I)(F)F)(F)F)(C(C(F)(F)I)(F)F)(F)F
375-80-4
C6F12I2
553.856
MFCD00042264
JOQDDLBOAIKFQX-UHFFFAOYSA-N
1,6-diiodoperfluorohexane,1,6-diiodododecafluorohexane,dodecafluoro-1,6-diiodohexane,1,6-diiododecafluorohexane,perfluoro-1,6-diiodohexane,hexane, 1,1,2,2,3,3,4,4,5,5,6,6-dodecafluoro-1,6-diiodo,acmc-209iu0,1,1,2,2,3,3,4,4,5,5,6,6-dodecafluoro-1,6-diiodo-hexane,hexane,1,1,2,2,3,3,4,4,5,5,6,6-dodecafluoro-1,6-diiodo
78994
1,1,2,2,3,3,4,4,5,5,6,6-dodecafluoro-1,6-diiodohexane
C(C(C(C(F)(F)I)(F)F)(F)F)(C(C(F)(F)I)(F)F)(F)F
Applications
It is employed in the synthesis and characterization of fluorinated telomers containing vinylidene fluoride and hexafluoropropene.

Notes
Light sensitive. Protect from light. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Quantity 25g
Melting Point 25°C
Boiling Point 115°C (100mmHg)
Beilstein 1822186
Formula Weight 553.86
Percent Purity 97%
Chemical Name or Material 1,6-Diiodoperfluorohexane