Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,6-Diisocyanatohexane 99+%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
1kg
Packaging:
Glass bottle
822-06-0
C8H12N2O2
168.19
MFCD00002047
RRAMGCGOFNQTLD-UHFFFAOYSA-N
hexamethylene diisocyanate,1,6-hexamethylene diisocyanate,hmdi,hexane, 1,6-diisocyanato,hexane 1,6-diisocyanate,1,6-hexylene diisocyanate,hexamethylene-1,6-diisocyanate,1,6-hexanediol diisocyanate,hdi,isocyanic acid, hexamethylene ester
13192
CHEBI:53578
1,6-diisocyanatohexane
C(CCCN=C=O)CCN=C=O
822-06-0
C8H12N2O2
168.19
MFCD00002047
RRAMGCGOFNQTLD-UHFFFAOYSA-N
hexamethylene diisocyanate,1,6-hexamethylene diisocyanate,hmdi,hexane, 1,6-diisocyanato,hexane 1,6-diisocyanate,1,6-hexylene diisocyanate,hexamethylene-1,6-diisocyanate,1,6-hexanediol diisocyanate,hdi,isocyanic acid, hexamethylene ester
13192
CHEBI:53578
1,6-diisocyanatohexane
C(CCCN=C=O)CCN=C=O
Melting Point -67°C
Specific Gravity 1.04
Flash Point 130°C
Packaging Glass bottle
Boiling Point 255.0°C
Infrared Spectrum Authentic
Density 1.0400g/mL
Quantity 100g
Color Colorless to Yellow
Refractive Index 1.4510 to 1.4530
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Linear Formula OCN(CH2)6NCO
Beilstein 04, II, 711
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in benzene,acetone,toluene,,ethyl acetate,chlorobenzene,very soluble in ether
Formula Weight 168.19
Physical Form Liquid
Percent Purity ≥99%
Chemical Name or Material 1, 6-Diisocyanatohexane, 99+%
Show More Show Less

GHS H StatementToxic if inhaled.Causes serious eye irritation.May cause respiratory irritation.Causes skin irritation.May cause allergy or asthma symptoms or breathing difficulties if inhaled.May cause an allerg

GHS P StatementIF SWALLOWED: rinse mouth.Do NOT induce vomiting.IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.Call a POISON CENTER or doctor/physician.IF ON SKIN: Wash with plenty of soap and water.If skin irritation or rash occurs: Get medical advice/attention.Wear protective gloves/protective clothing/eye protection/face protection.If skin irritation occurs: Get medical advice/attention.

;GHS P StatementIF SWALLOWED: rinse mouth. Do NOT induce vomiting.IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.Call a POISON CENTER or doctor/physician.IF ON SKIN: Wash with plenty of soap and wat

EINECSNumber : 212-485-8

RTECSNumber : MO1740000

TSCA : TSCA