Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,6-Dimethoxyhexane, 97%

Click to view available options
Quantity:
5g
25g
100g
13179-98-1
C8H18O2
146.23
MFCD00142569
GZHQUHIMIVEQFV-UHFFFAOYSA-N
hexane,1,6-dimethoxy,hexamethylene glycol dimethyl ether,hexane, 1,6-dimethoxy,acmc-1c0t4,1,6-dimethoxyhexane
542359
1,6-dimethoxyhexane
COCCCCCCOC
13179-98-1
C8H18O2
146.23
MFCD00142569
GZHQUHIMIVEQFV-UHFFFAOYSA-N
hexane,1,6-dimethoxy,hexamethylene glycol dimethyl ether,hexane, 1,6-dimethoxy,acmc-1c0t4,1,6-dimethoxyhexane
542359
1,6-dimethoxyhexane
COCCCCCCOC
Quantity 25g
Boiling Point 182°C to 183°C
Flash Point 69°C (157°F)
Formula Weight 146.23
Percent Purity 97%
Chemical Name or Material 1,6-Dimethoxyhexane

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning