Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,6-Diphenyl-1,3,5-hexatriene 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
1720-32-7
C18H16
232.32
MFCD00004793
BOBLSBAZCVBABY-WPWUJOAOSA-N
1,6-diphenyl-1,3,5-hexatriene,1,6-diphenylhexatriene,diphenylhexatriene,dicinnamyl,1,6-diphenylhexa-1,3,5-triene,benzene, 1,1'-1,3,5-hexatriene-1,6-diyl bis,1,3,5-hexatriene, 1,6-diphenyl,1e,3e,5e-6-phenylhexa-1,3,5-trienyl benzene,1,1'-1e,3e,5e-hexa-1,3,5-triene-1,6-diyldibenzene
5376733
CHEBI:51594
[(1E,3E,5E)-6-phenylhexa-1,3,5-trienyl]benzene
C1=CC=C(C=C1)C=CC=CC=CC2=CC=CC=C2
1720-32-7
C18H16
232.32
MFCD00004793
BOBLSBAZCVBABY-WPWUJOAOSA-N
1,6-diphenyl-1,3,5-hexatriene,1,6-diphenylhexatriene,diphenylhexatriene,dicinnamyl,1,6-diphenylhexa-1,3,5-triene,benzene, 1,1'-1,3,5-hexatriene-1,6-diyl bis,1,3,5-hexatriene, 1,6-diphenyl,1e,3e,5e-6-phenylhexa-1,3,5-trienyl benzene,1,1'-1e,3e,5e-hexa-1,3,5-triene-1,6-diyldibenzene
5376733
CHEBI:51594
[(1E,3E,5E)-6-phenylhexa-1,3,5-trienyl]benzene
C1=CC=C(C=C1)C=CC=CC=CC2=CC=CC=C2
Infrared Spectrum Authentic
Color Yellow
Melting Point 199°C to 203°C
Quantity 5g
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula C6H5CH=CHCH=CHCH=CHC6H5
Beilstein 05, 691
Formula Weight 232.32
Physical Form Crystalline Powder or Crystals
Percent Purity ≥97.5%
Chemical Name or Material 1,6-Diphenyl-1,3,5-hexatriene, 98%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 217-011-3

TSCA : TSCA