Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,6-Heptadien-4-ol 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
2883-45-6
C7H12O
112.17
MFCD00008663
UTGFOWQYZKTZTN-UHFFFAOYSA-N
1,6-heptadien-4-ol,1,6-heptadiene-4-ol,diallylcarbinol,pubchem13740,4-hepta-1,6-dienol,acmc-1cei5,intermediates-zcf02285,4-hydroxy-1,6-heptadiene,4-01-00-02249 beilstein handbook reference
17902
hepta-1,6-dien-4-ol
C=CCC(CC=C)O
2883-45-6
C7H12O
112.17
MFCD00008663
UTGFOWQYZKTZTN-UHFFFAOYSA-N
1,6-heptadien-4-ol,1,6-heptadiene-4-ol,diallylcarbinol,pubchem13740,4-hepta-1,6-dienol,acmc-1cei5,intermediates-zcf02285,4-hydroxy-1,6-heptadiene,4-01-00-02249 beilstein handbook reference
17902
hepta-1,6-dien-4-ol
C=CCC(CC=C)O
Boiling Point 151°C
Flash Point 40°C
Specific Gravity 0.86
Density 0.8600g/mL
Refractive Index 1.4490 to 1.453
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 25g
Color Colorless to Yellow
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula H2C=CHCH2CH(OH)CH2CH=CH2
Beilstein 01, 455
Formula Weight 112.17
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material 1, 6-Heptadien-4-ol, 97%
Show More Show Less

GHS H StatementFlammable liquid and vapor.Causes skin irritation.Causes serious eye irritation.May cause respiratory irritation.

GHS P StatementKeep away from heat/sparks/open flames/hot surfaces.- No smoking.IF ON SKIN (or hair): Take off immediately all contaminated clothing.Rinse skin with water/shower.Wash face,hands and any exposed skin thoroughly after handling.

;GHS P StatementKeep away from heat/sparks/open flames/hot surfaces.- No smoking.IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.Wash face,hands and any exposed skin thoroughly after handli

EINECSNumber : 220-742-0

RTECSNumber : MI5450000

TSCA : TSCA