Alfa Aesar™ 1,6-Heptadiyne, 97%

Manufacturer: Alfa Aesar™ A1131822

Quantity 100g

 View more versions of this product

Catalog No. AAA1131822

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2396-63-6
    Molecular Formula C7H8
    Formula Weight 92.14
    MDL Number MFCD00014925
    UN Number UN3295
    Beilstein 1731756
    Chemical Name or Material 1,6-Heptadiyne

    Specifications

    Formula Weight 92.14
    Health Hazard 3 GHS P Statement
    P210-P243-P301+P330+P331-P315
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Take precautionary measures against static discharge.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Get immediate medical advice/attention.
    CAS 2396-63-6
    RTECS Number MI5600000
    TSCA TSCA
    Beilstein 1731756
    Recommended Storage 2°C to 8°C
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
    Flash Point 9°C (48°F)
    Molecular Formula C7H8
    Health Hazard 1 Danger
    Odor Recognizable
    Boiling Point 109° to 111°C
    MDL Number MFCD00014925
    UN Number UN3295
    Health Hazard 2 GHS H Statement
    H225-H304
    Highly flammable liquid and vapor.
    May be fatal if swallowed and enters airways.
    Quantity 100g
    Chemical Name or Material 1,6-Heptadiyne
    Assay 97%

    Description

    Applications
    1,6-Heptadiyne may be used in the preparation of free-standing polymer films of poly(1,6-heptadiyne). Polymerization of 1,6-heptadiyne using an insoluble Zeigler-Natta catalyst affords soluble polymer having six-membered ring containing polyene. Ruthenium(II)-catalyzed reaction of a substituted 1,6-heptadiyne with norbornene affords a tandem [2+ 2+ 2]/[4+2] cycloaddition product and a [2+ 2+2] cycloadduct.

    Solubility
    Insoluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Haevy metal powder, heat. Refrigerate at (+4°C).

    GHS H Statement
    H225-H304
    Highly flammable liquid and vapor.
    May be fatal if swallowed and enters airways.

    2°C to 8°C

    GHS P Statement
    P210-P243-P301+P330+P331-P315
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Take precautionary measures against static discharge.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Get immediate medical advice/attention.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit