Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,6-Hexanediol, 97%

Manufacturer:  Alfa Aesar A1243936

Quantity 500g

 View more versions of this product

Catalog No. AAA1243936


  Check Availability
Add to cart

Specifications

Description

Applications
1,6-Hexanediol is used in polymer synthesis such as polyester, polyurethane and nylon. It is used as an intermediate to adhesives, acrylics and dyestuffs. Further, it is employed in gasoline refining and pharmaceutical production.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with acid chlorides, acid anhydrides, oxidizing agents, chloroformates and reducing agents.
Description & Specifications

Chemical Identifiers

629-11-8
118.18
1633461
1, 6-Hexanediol
HO(CH2)6OH
MFCD00002985
14,4690

Specifications

500g
1.120
HO(CH2)6OH
MFCD00002985
14,4690
97%
629-11-8
97%
118.18
1633461
1, 6-Hexanediol

Safety and Handling

SDS & Safety Documents

RTECSNumber : MO2100000

TSCA : TSCA

storageNote1 : Hygroscopic

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit