Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,6-Naphthyridine-2-carboxylic acid, 98%, Acros Organics™

Click to view available options
Quantity:
250mg
1g
197507-59-8
C9H6N2O2
174.16
OZZMWXQJCJUCEJ-UHFFFAOYSA-N
1,6-naphthyridine-2-carboxylicacid,1,6 naphthyridine-2-carboxylic acid,peakdale1_000097,2-1,6 naphthyridinecarboxylic acid,2-carboxy-1,6-naphthyridine,pyridino 4,3-b pyridine-2-carboxylic acid
735908
1,6-naphthyridine-2-carboxylic acid
C1=CC(=NC2=C1C=NC=C2)C(=O)O
197507-59-8
C9H6N2O2
174.16
OZZMWXQJCJUCEJ-UHFFFAOYSA-N
1,6-naphthyridine-2-carboxylicacid,1,6 naphthyridine-2-carboxylic acid,peakdale1_000097,2-1,6 naphthyridinecarboxylic acid,2-carboxy-1,6-naphthyridine,pyridino 4,3-b pyridine-2-carboxylic acid
735908
1,6-naphthyridine-2-carboxylic acid
C1=CC(=NC2=C1C=NC=C2)C(=O)O
Infrared Spectrum Authentic
Quantity 250mg
Melting Point 290.0°C
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 174.16
Percent Purity 98%
Chemical Name or Material 1,6-Naphthyridine-2-carboxylic acid

GHS H Statement Causes skin irritation. Causes serious eye irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning