Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,7-Diacetoxyheptane 95.0+%, TCI America™

Click to view available options
Quantity:
10g
52819-34-8
C11H20O4
216.277
MFCD00191657
MABSJAOVZMJMDT-UHFFFAOYSA-N
Heptamethylene Diacetate, 1,7-Heptanediol Diacetate
537565
7-acetyloxyheptyl acetate
CC(=O)OCCCCCCCOC(=O)C
52819-34-8
C11H20O4
216.277
MFCD00191657
MABSJAOVZMJMDT-UHFFFAOYSA-N
Heptamethylene Diacetate, 1,7-Heptanediol Diacetate
537565
7-acetyloxyheptyl acetate
CC(=O)OCCCCCCCOC(=O)C
Quantity 10g
Color Yellow
Boiling Point 270°C
Formula Weight 216.28
Physical Form Clear Liquid at 20°C
Percent Purity ≥95.0% (GC)
Chemical Name or Material 1,7-Diacetoxyheptane