Alfa Aesar™ 1,7-Dibenzyl-1,4,7,10-tetraazacyclododecane

Manufacturer: Alfa Aesar™ H26517MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH26517MD

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 156970-79-5
    Molecular Formula C22H32N4
    Formula Weight 352.52
    MDL Number MFCD09263309
    Synonym N(1),N(7)-Dibenzylcyclen
    Chemical Name or Material 1,7-Dibenzyl-1,4,7,10-tetraazacyclododecane

    Specifications

    Formula Weight 352.52
    Quantity 250mg
    Molecular Formula C22H32N4
    MDL Number MFCD09263309
    CAS 156970-79-5
    Synonym N(1),N(7)-Dibenzylcyclen
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 1,7-Dibenzyl-1,4,7,10-tetraazacyclododecane

    Description

    Applications
    1,7-Dibenzyl-1,4,7,10-tetraazacyclododecane is used as a pharmaceutical intermediate.

    Solubility
    Soluble in acetone, acetonitrile, ethyl acetate, chloroform, cyclohexane, dichloromethane, ether, methanol, pentane and toluene. Insoluble in water.

    Notes
    Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Store under dry inert gas. It is hygroscopic in nature. Incompatible with oxidizing agents. Protect from humidity and water.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit