Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,7-Dioxaspiro(5.5)undecane, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
180-84-7
C9H16O2
156.225
MFCD00011578
GBBVHDGKDQAEOT-UHFFFAOYSA-N
1,7-dioxaspiro 5.5 undecane,olean,unii-khi3qeq4zj,1,7-dioxaspiro 5,5 undecane,khi3qeq4zj,r-1,7-dioxaspiro 5.5 undecane,s-1,7-dioxaspiro 5.5 undecane,olive-fly ketal,eco-trap
67437
1,7-dioxaspiro[5.5]undecane
C1CCOC2(C1)CCCCO2
180-84-7
C9H16O2
156.225
MFCD00011578
GBBVHDGKDQAEOT-UHFFFAOYSA-N
1,7-dioxaspiro 5.5 undecane,olean,unii-khi3qeq4zj,1,7-dioxaspiro 5,5 undecane,khi3qeq4zj,r-1,7-dioxaspiro 5.5 undecane,s-1,7-dioxaspiro 5.5 undecane,olive-fly ketal,eco-trap
67437
1,7-dioxaspiro[5.5]undecane
C1CCOC2(C1)CCCCO2
Quantity 1g
Density 1.0200g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Color Colorless
Refractive Index 1.4620 to 1.4640 (20°C, 589nm)
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 156.23
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material 1, 7-Dioxaspiro(5.5)undecane, 97%
Show More Show Less