Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,7-Heptanediol, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
10g
Packaging:
Glass bottle
629-30-1
C7H16O2
132.2
MFCD00002987
SXCBDZAEHILGLM-UHFFFAOYSA-N
1,7-heptanediol,1,7-dihydroxyheptane,heptamethylene glycol,alpha,omega-heptanediol,.alpha.,.omega.-heptanediol,a,?-heptanediol,1,7-heptandiol,.omega.-heptanediol,pubchem11489,acmc-1bsor
12381
heptane-1,7-diol
C(CCCO)CCCO
629-30-1
C7H16O2
132.2
MFCD00002987
SXCBDZAEHILGLM-UHFFFAOYSA-N
1,7-heptanediol,1,7-dihydroxyheptane,heptamethylene glycol,alpha,omega-heptanediol,.alpha.,.omega.-heptanediol,a,?-heptanediol,1,7-heptandiol,.omega.-heptanediol,pubchem11489,acmc-1bsor
12381
heptane-1,7-diol
C(CCCO)CCCO
Flash Point >110°C
Specific Gravity 0.951
Boiling Point 259°C
Melting Point 17°C to 19°C
Density 0.9510g/mL
Refractive Index 1.4535 to 1.4565
Quantity 1g
Infrared Spectrum Authentic
Color Colorless to Yellow
Packaging Glass bottle
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Linear Formula HO(CH2)7OH
Beilstein 01, 489
Formula Weight 132.2
Physical Form Liquid After Melting
Percent Purity 95%
Chemical Name or Material 1, 7-Heptanediol, 95%
Show More Show Less

EINECSNumber : 211-085-0

RTECSNumber : MI9804000

TSCA : TSCA