Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,7-Octadiyne, 98%, Acros Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
871-84-1
C8H10
106.16
MFCD00008580
DSOJWVLXZNRKCS-UHFFFAOYSA-N
1,7-octadiyne,1,7 octadiyne,oct-1,7-diyne,acmc-209qii,3-thia-1,5-pentanediamine,1,7-octadiyne 10g,1,7-octadiyne 6ci, 7ci, 8ci, 9ci
70099
octa-1,7-diyne
C#CCCCCC#C
871-84-1
C8H10
106.16
MFCD00008580
DSOJWVLXZNRKCS-UHFFFAOYSA-N
1,7-octadiyne,1,7 octadiyne,oct-1,7-diyne,acmc-209qii,3-thia-1,5-pentanediamine,1,7-octadiyne 10g,1,7-octadiyne 6ci, 7ci, 8ci, 9ci
70099
octa-1,7-diyne
C#CCCCCC#C
Refractive Index 1.4454 to 1.4474
Infrared Spectrum Authentic
Packaging Glass bottle
Specific Gravity 0.8
Flash Point 23°C
Boiling Point 135.0°C to 136.0°C
Quantity 10g
Color Colorless to Yellow
Density 0.8000g/mL
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula HC≡C(CH2)4C≡CH
Beilstein 01, II, 248
Formula Weight 106.16
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 1, 7-Octadiyne, 98%
Show More Show Less

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 212-815-0

TSCA : TSCA