Alfa Aesar™ 1,7-Phenanthroline

Manufacturer: Alfa Aesar™ 3090906

Quantity 5g

 View more versions of this product

Catalog No. AA3090906

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 230-46-6
    Formula Weight 180.21
    MDL Number MFCD00004971
    UN Number UN2811
    Chemical Name or Material 1,7-Phenanthroline

    Specifications

    Formula Weight 180.21
    Health Hazard 3 GHS P Statement
    P261-P280-P281-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number SF8290000
    Odor Odorless
    Physical Form Crystalline
    MDL Number MFCD00004971
    UN Number UN2811
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    CAS 230-46-6
    Melting Point 77° to 81°C
    Quantity 5g
    Health Hazard 2 GHS H Statement
    H331-H341-H302-H312
    Toxic if inhaled.
    Suspected of causing genetic defects.
    Harmful if swallowed.
    Harmful in contact with skin.
    Chemical Name or Material 1,7-Phenanthroline

    GHS H Statement
    H331-H341-H302-H312
    Toxic if inhaled.
    Suspected of causing genetic defects.
    Harmful if swallowed.
    Harmful in contact with skin.

    GHS P Statement
    P261-P280-P281-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit