Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,8-Diaminonaphthalene, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
2.5kg
Packaging:
Glass Bottle
479-27-6
C10H10N2
158.2
MFCD00004033
YFOOEYJGMMJJLS-UHFFFAOYSA-N
1,8-diaminonaphthalene,1,8-naphthalenediamine,1,8-naphthylenediamine,unii-ika7029yh9,ccris 8398,1,8-diaminonapthalene,1,8-diamino naphthalene,1,8-naphthalene diamine,acmc-209kb1,ksc236e9t
68067
naphthalene-1,8-diamine
C1=CC2=C(C(=C1)N)C(=CC=C2)N
479-27-6
C10H10N2
158.2
MFCD00004033
YFOOEYJGMMJJLS-UHFFFAOYSA-N
1,8-diaminonaphthalene,1,8-naphthalenediamine,1,8-naphthylenediamine,unii-ika7029yh9,ccris 8398,1,8-diaminonapthalene,1,8-diamino naphthalene,1,8-naphthalene diamine,acmc-209kb1,ksc236e9t
68067
naphthalene-1,8-diamine
C1=CC2=C(C(=C1)N)C(=CC=C2)N
Packaging Glass Bottle
Flash Point 195°C
Quantity 2.5kg
Infrared Spectrum Authentic
Boiling Point 205.0°C (12.0 mmHg)
Color Brown or Gray to Gray-Red
Melting Point 63.0°C to 66.0°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula C10H6(NH2)2
Beilstein 13, 205
Merck Index 15, 6457
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol and ether, slightly soluble in chloroform
Formula Weight 158.2
Physical Form Chunks or Crystalline Mass
Percent Purity 97%
Chemical Name or Material 1, 8-Diaminonaphthalene, 97%
Show More Show Less

GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If s

GHS Signal Word: Warning

EINECSNumber : 207-529-8