Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,8-Diaminonaphthalene, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
2.5kg
Packaging:
Glass bottle
479-27-6
C10H10N2
158.2
MFCD00004033
YFOOEYJGMMJJLS-UHFFFAOYSA-N
1,8-diaminonaphthalene,1,8-naphthalenediamine,1,8-naphthylenediamine,unii-ika7029yh9,ccris 8398,1,8-diaminonapthalene,1,8-diamino naphthalene,1,8-naphthalene diamine,acmc-209kb1,ksc236e9t
68067
naphthalene-1,8-diamine
C1=CC2=C(C(=C1)N)C(=CC=C2)N
479-27-6
C10H10N2
158.2
MFCD00004033
YFOOEYJGMMJJLS-UHFFFAOYSA-N
1,8-diaminonaphthalene,1,8-naphthalenediamine,1,8-naphthylenediamine,unii-ika7029yh9,ccris 8398,1,8-diaminonapthalene,1,8-diamino naphthalene,1,8-naphthalene diamine,acmc-209kb1,ksc236e9t
68067
naphthalene-1,8-diamine
C1=CC2=C(C(=C1)N)C(=CC=C2)N
Flash Point 195°C
Packaging Glass bottle
Color Brown or Gray to Gray-Red
Melting Point 63.0°C to 66.0°C
Quantity 500g
Infrared Spectrum Authentic
Boiling Point 205.0°C (12.0 mmHg)
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula C10H6(NH2)2
Beilstein 13, 205
Merck Index 15, 6457
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol and ether, slightly soluble in chloroform
Formula Weight 158.2
Physical Form Chunks or Crystalline Mass
Percent Purity 97%
Chemical Name or Material 1, 8-Diaminonaphthalene, 97%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Suspected of causing cancer.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Wear eye protection/face protection.
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.