Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,8-Dichloroanthraquinone, 96%

Click to view available options
Quantity:
25g
100g
500g
82-43-9
C14H6Cl2O2
277.1
MFCD00001191
VBQNYYXVDQUKIU-UHFFFAOYSA-N
1,8-dichloroanthraquinone,9,10-anthracenedione, 1,8-dichloro,1,8-dichloranthrachinon,1,8-dichloro-9,10-anthraquinone,anthraquinone, 1,8-dichloro,1,8-dichloranthrachinon czech,acmc-1bmbx,9, 1,8-dichloro,anthraquinone,8-dichloro
6708
1,8-dichloroanthracene-9,10-dione
C1=CC2=C(C(=C1)Cl)C(=O)C3=C(C2=O)C=CC=C3Cl
82-43-9
C14H6Cl2O2
277.1
MFCD00001191
VBQNYYXVDQUKIU-UHFFFAOYSA-N
1,8-dichloroanthraquinone,9,10-anthracenedione, 1,8-dichloro,1,8-dichloranthrachinon,1,8-dichloro-9,10-anthraquinone,anthraquinone, 1,8-dichloro,1,8-dichloranthrachinon czech,acmc-1bmbx,9, 1,8-dichloro,anthraquinone,8-dichloro
6708
1,8-dichloroanthracene-9,10-dione
C1=CC2=C(C(=C1)Cl)C(=O)C3=C(C2=O)C=CC=C3Cl
Applications
1,8-Dichloroanthraquinone is a reagent in the preparation of anthraquinones, which has antitumor activities. It is used as primary and secondary intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agent.
Quantity 100g
Melting Point 201°C to 203°C
Formula Weight 277.11
Percent Purity 96%
Chemical Name or Material 1,8-Dichloroanthraquinone

GHS H Statement
H319
Causes serious eye irritation.

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF eye irritation persists: Get medical advice/attention.

Warning

RTECSNumber : CB6496000

TSCA : TSCA