Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,8-dimethylnaphthalene, 98%, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
Packaging:
Glass bottle
569-41-5
C12H12
156.228
MFCD00004040
XAABPYINPXYOLM-UHFFFAOYSA-N
naphthalene, 1,8-dimethyl,unii-22p9fw1l76,1,8-dmn,1,8-dimethyl-naphthalene,naphthalene,1,8-dimethyl,1,8-dimethylnaphthalene,1,8-dimethylnaphthalene, analytical standard,10 microg/ml in cyclohexane
11287
CHEBI:48610
1,8-dimethylnaphthalene
CC1=CC=CC2=C1C(=CC=C2)C
569-41-5
C12H12
156.228
MFCD00004040
XAABPYINPXYOLM-UHFFFAOYSA-N
naphthalene, 1,8-dimethyl,unii-22p9fw1l76,1,8-dmn,1,8-dimethyl-naphthalene,naphthalene,1,8-dimethyl,1,8-dimethylnaphthalene,1,8-dimethylnaphthalene, analytical standard,10 microg/ml in cyclohexane
11287
CHEBI:48610
1,8-dimethylnaphthalene
CC1=CC=CC2=C1C(=CC=C2)C
Melting Point 58°C to 62°C
Packaging Glass bottle
Quantity 250mg
Color Beige-Orange to White
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula C10H6(CH3)2
Formula Weight 156.23
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 1, 8-Dimethylnaphthalene
Show More Show Less

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.