Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,8-Naphthalaldehydic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
5811-87-0
C12H8O3
200.193
MFCD00134159
HLHDIWAOQIRETC-UHFFFAOYSA-N
8-formyl-1-naphthoic acid,1,8-naphthalaldehydic acid,naphthaldehydic acid,8-formyl-1-naphthalenecarboxylic acid,naphthalaldehydic acid,1-naphthalenecarboxylic acid, 8-formyl,peri-naphthalaldehydic acid,8-napthalic acid,1,8-naphthaldehydic acid
79881
8-formylnaphthalene-1-carboxylic acid
C1=CC2=C(C(=C1)C=O)C(=CC=C2)C(=O)O
5811-87-0
C12H8O3
200.193
MFCD00134159
HLHDIWAOQIRETC-UHFFFAOYSA-N
8-formyl-1-naphthoic acid,1,8-naphthalaldehydic acid,naphthaldehydic acid,8-formyl-1-naphthalenecarboxylic acid,naphthalaldehydic acid,1-naphthalenecarboxylic acid, 8-formyl,peri-naphthalaldehydic acid,8-napthalic acid,1,8-naphthaldehydic acid
79881
8-formylnaphthalene-1-carboxylic acid
C1=CC2=C(C(=C1)C=O)C(=CC=C2)C(=O)O
Color Cream
Infrared Spectrum Authentic
Melting Point 160°C to 167°C
Quantity 1g
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 200.19
Physical Form Powder
Percent Purity ≥96.0%
Chemical Name or Material 1, 8-Naphthalaldehydic acid
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.