Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,8-Octanediol, 98+%, Alfa Aesar™

Click to view available options
Quantity:
25g
100g
500g
629-41-4
C8H18O2
146.23
MFCD00002989
OEIJHBUUFURJLI-UHFFFAOYSA-N
1,8-octanediol,octamethylene glycol,1,8-dihydroxyoctane,octan-1,8-diol,1,8-octandiol,1, 8-octandiol,octane-1,8-diol;,acmc-209nan,ksc358i9p
69420
CHEBI:44630
octane-1,8-diol
C(CCCCO)CCCO
629-41-4
C8H18O2
146.23
MFCD00002989
OEIJHBUUFURJLI-UHFFFAOYSA-N
1,8-octanediol,octamethylene glycol,1,8-dihydroxyoctane,octan-1,8-diol,1,8-octandiol,1, 8-octandiol,octane-1,8-diol;,acmc-209nan,ksc358i9p
69420
CHEBI:44630
octane-1,8-diol
C(CCCCO)CCCO
Applications
1,8-Octanediol is used in cosmetics, perfumes, ink, essence and in UV coating. It acts as a precursor in the synthesis of polymer and in the manufacture of pharmaceuticals.

Solubility
Soluble in water and methanol.

Notes
Incompatible with strong oxidizing agents, reducing agents, acid chlorides acid, anhydrides and chloroformates.
Melting Point 58°C to 61°C
Quantity 500g
Flash Point 148°C (298°F)
Boiling Point 171°C to 173°C (20mmHg)
Assay Percent Range ≥98%
Linear Formula HO(CH2)8OH
Beilstein 1633499
Solubility Information Soluble in water and methanol.
Formula Weight 146.23
Percent Purity ≥98%
Chemical Name or Material 1,8-Octanediol
Show More Show Less

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 211-090-8

TSCA : No

Recommended Storage : Ambient temperatures