Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,8-Octanedithiol, 98%

Click to view available options
Quantity:
1g
5g
25g
1191-62-4
C8H18S2
178.352
MFCD00003574
PGTWZHXOSWQKCY-UHFFFAOYSA-N
1,8-octanedithiol,1,8-dimercaptooctane,octamethylene dimercaptan,1,8-octamethylenedithiol,unii-kh3w3161hs,fema no. 3514,1,8-octane dithiol,nanothinks tm thio8,acmc-209a1j,dsstox_cid_27474
14493
octane-1,8-dithiol
C(CCCCS)CCCS
1191-62-4
C8H18S2
178.352
MFCD00003574
PGTWZHXOSWQKCY-UHFFFAOYSA-N
1,8-octanedithiol,1,8-dimercaptooctane,octamethylene dimercaptan,1,8-octamethylenedithiol,unii-kh3w3161hs,fema no. 3514,1,8-octane dithiol,nanothinks tm thio8,acmc-209a1j,dsstox_cid_27474
14493
octane-1,8-dithiol
C(CCCCS)CCCS
Odor Unpleasant
Flash Point 113°C (235°F)
Boiling Point 269°C to 270°C
Quantity 25g
Beilstein 1735431
Formula Weight 178.36
Percent Purity 98%
Chemical Name or Material 1,8-Octanedithiol

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : RG9610000