Learn More
SWAGELOK TEXAS MID COAST 1/8 T. X 1/4 MPT
Supplier:  SWAGELOK TEXAS MID COAST M20014
Encompass
Alloy 400/R-405 Swagelok Tube Fitting, Male Connector, 1/8 in. Tube OD x 1/4 in. Male NPT
Catalog No. NC2306706
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.