Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,9-Dichlorononane, 98%

Click to view available options
Quantity:
5g
25g
100g
821-99-8
C9H18Cl2
197.143
MFCD00001024
JMGRNJZUQCEJDB-UHFFFAOYSA-N
nonane, 1,9-dichloro,1,9-dichlornonane,nonane,9-dichloro,ksc449k9f,1,9-dichlornonan;1,9-dichloro-nonan;nonane, 1,9-dichloro-;nonane,1,9-dichloro-;
13191
1,9-dichlorononane
C(CCCCCl)CCCCCl
821-99-8
C9H18Cl2
197.143
MFCD00001024
JMGRNJZUQCEJDB-UHFFFAOYSA-N
nonane, 1,9-dichloro,1,9-dichlornonane,nonane,9-dichloro,ksc449k9f,1,9-dichlornonan;1,9-dichloro-nonan;nonane, 1,9-dichloro-;nonane,1,9-dichloro-;
13191
1,9-dichlorononane
C(CCCCCl)CCCCCl
Applications
It is used as pharmaceutical intermediate. Many organisms use 1,9-dichlorononane as a sole carbon source. Bacteria on incubation of sewage with 1,9-dichlorononane increased its subsequent capacity to dehalogenate.

Solubility
Insoluble in water.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Refractive Index 1.4590
Boiling Point 258°C to 262°C
Quantity 25g
Color Colorless
Density 1.09g/mL
Linear Formula Cl(CH2)9Cl
Beilstein 1698729
Formula Weight 197.14
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 1,9-Dichlorononane
Show More Show Less