Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,9-Dichlorononane, 98%

Click to view available options
Quantity:
5g
25g
100g
821-99-8
C9H18Cl2
197.143
MFCD00001024
JMGRNJZUQCEJDB-UHFFFAOYSA-N
nonane, 1,9-dichloro,1,9-dichlornonane,nonane,9-dichloro,ksc449k9f,1,9-dichlornonan;1,9-dichloro-nonan;nonane, 1,9-dichloro-;nonane,1,9-dichloro-;
13191
1,9-dichlorononane
C(CCCCCl)CCCCCl
821-99-8
C9H18Cl2
197.143
MFCD00001024
JMGRNJZUQCEJDB-UHFFFAOYSA-N
nonane, 1,9-dichloro,1,9-dichlornonane,nonane,9-dichloro,ksc449k9f,1,9-dichlornonan;1,9-dichloro-nonan;nonane, 1,9-dichloro-;nonane,1,9-dichloro-;
13191
1,9-dichlorononane
C(CCCCCl)CCCCCl
Applications
It is used as pharmaceutical intermediate. Many organisms use 1,9-dichlorononane as a sole carbon source. Bacteria on incubation of sewage with 1,9-dichlorononane increased its subsequent capacity to dehalogenate.

Solubility
Insoluble in water.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Color Colorless
Density 1.09g/mL
Quantity 100g
Refractive Index 1.4590
Boiling Point 258°C to 262°C
Linear Formula Cl(CH2)9Cl
Beilstein 1698729
Formula Weight 197.14
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 1,9-Dichlorononane
Show More Show Less