Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Acetonylpyridinium Chloride 99.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
42508-60-1
C8H10ClNO
171.624
MFCD00031949
JHYYWKUENNZTMD-UHFFFAOYSA-M
1-acetonylpyridinium chloride,n-acetonylpyridinium chloride,1-2-oxopropyl pyridinium chloride,1-2-oxopropyl pyridin-1-ium chloride,acmc-209jpw,1-acetonylpyridiniumchloride,1-pyridin-1-ium-1-ylpropan-2-one chloride,1-2-oxopropyl-2h-pyridin-2-ylium chloride
2734121
1-pyridin-1-ium-1-ylpropan-2-one;chloride
CC(=O)C[N+]1=CC=CC=C1.[Cl-]
42508-60-1
C8H10ClNO
171.624
MFCD00031949
JHYYWKUENNZTMD-UHFFFAOYSA-M
1-acetonylpyridinium chloride,n-acetonylpyridinium chloride,1-2-oxopropyl pyridinium chloride,1-2-oxopropyl pyridin-1-ium chloride,acmc-209jpw,1-acetonylpyridiniumchloride,1-pyridin-1-ium-1-ylpropan-2-one chloride,1-2-oxopropyl-2h-pyridin-2-ylium chloride
2734121
1-pyridin-1-ium-1-ylpropan-2-one;chloride
CC(=O)C[N+]1=CC=CC=C1.[Cl-]
Color White-Yellow
Melting Point 207°C
Quantity 25g
Formula Weight 171.62
Physical Form Crystal-Powder at 20°C
Percent Purity ≥99.0% (HPLC,T)
Chemical Name or Material 1-Acetonylpyridinium Chloride