Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Acetyl-4-fluoronaphthalene, 97%

Click to view available options
Quantity:
1g
5g
25g
316-68-7
C12H9FO
188.201
MFCD00134475
GAMOBQXCYKWQLT-UHFFFAOYSA-N
1-4-fluoronaphthalen-1-yl ethanone,1-acetyl-4-fluoronaphthalene,1-4-fluoro-1-naphthyl ethanone,4-fluoro-1-acetonaphthone,ethanone, 1-4-fluoro-1-naphthalenyl,1-4-fluoro-1-naphthyl ethan-1-one,4'-fluoro-1'-acetonaphthone,1-4-fluoronaphthalen-1-yl ethan-1-one,4-fluoroacetonaphthone,4'-fluoro-1'acetonaphthone
67568
1-(4-fluoronaphthalen-1-yl)ethanone
CC(=O)C1=CC=C(C2=CC=CC=C21)F
316-68-7
C12H9FO
188.201
MFCD00134475
GAMOBQXCYKWQLT-UHFFFAOYSA-N
1-4-fluoronaphthalen-1-yl ethanone,1-acetyl-4-fluoronaphthalene,1-4-fluoro-1-naphthyl ethanone,4-fluoro-1-acetonaphthone,ethanone, 1-4-fluoro-1-naphthalenyl,1-4-fluoro-1-naphthyl ethan-1-one,4'-fluoro-1'-acetonaphthone,1-4-fluoronaphthalen-1-yl ethan-1-one,4-fluoroacetonaphthone,4'-fluoro-1'acetonaphthone
67568
1-(4-fluoronaphthalen-1-yl)ethanone
CC(=O)C1=CC=C(C2=CC=CC=C21)F
Quantity 5g
Boiling Point 164°C to 166°C (14mmHg)
Flash Point >110°C (230°F)
Formula Weight 188.2
Percent Purity 97%
Chemical Name or Material 1-Acetyl-4-fluoronaphthalene

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning