Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Acetyl-4-piperidinecarboxylic acid, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
50g
Packaging:
Glass bottle
25503-90-6
C8H13NO3
171.196
MFCD00023706
WFCLWJHOKCQYOQ-UHFFFAOYSA-N
1-acetyl-4-piperidinecarboxylic acid,1-acetylisonipecotic acid,1-acetyl-4-piperidine carboxylic acid,4-piperidinecarboxylic acid, 1-acetyl,n-acetyl-4-piperidinecarboxylic acid,1-acetyl-piperidine-4-carboxylic acid,maybridge1_003128,1-acetylpiperidine-4-carboxylic acid,,acetylisonipecotic acid,acmc-209gkl
117255
1-acetylpiperidine-4-carboxylic acid
CC(=O)N1CCC(CC1)C(=O)O
25503-90-6
C8H13NO3
171.196
MFCD00023706
WFCLWJHOKCQYOQ-UHFFFAOYSA-N
1-acetyl-4-piperidinecarboxylic acid,1-acetylisonipecotic acid,1-acetyl-4-piperidine carboxylic acid,4-piperidinecarboxylic acid, 1-acetyl,n-acetyl-4-piperidinecarboxylic acid,1-acetyl-piperidine-4-carboxylic acid,maybridge1_003128,1-acetylpiperidine-4-carboxylic acid,,acetylisonipecotic acid,acmc-209gkl
117255
1-acetylpiperidine-4-carboxylic acid
CC(=O)N1CCC(CC1)C(=O)O
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 180.0°C to 182.0°C
Color White to Beige
Quantity 5g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Solubility Information Solubility in water: 50g/L (20°C). Other solubilities: soluble in methanol, ethanol, benzene and toluene
Formula Weight 171.19
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material 1-Acetyl-4-piperidinecarboxylic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.