Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Acetyl-4-piperidone 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
32161-06-1
C7H11NO2
141.17
MFCD00006190
NNFOVLFUGLWWCL-UHFFFAOYSA-N
1-acetyl-4-piperidone,n-acetyl-4-piperidone,4-piperidinone, 1-acetyl,1-acetyl-4-piperidinone,1-acetyl-piperidin-4-one,1-acetyl-4-oxopiperidine,n-acetyl-4-piperidinone,n-acetyl4-piperidone,pubchem19615,1-acetylpiperid-4-one
122563
1-acetylpiperidin-4-one
CC(=O)N1CCC(=O)CC1
32161-06-1
C7H11NO2
141.17
MFCD00006190
NNFOVLFUGLWWCL-UHFFFAOYSA-N
1-acetyl-4-piperidone,n-acetyl-4-piperidone,4-piperidinone, 1-acetyl,1-acetyl-4-piperidinone,1-acetyl-piperidin-4-one,1-acetyl-4-oxopiperidine,n-acetyl-4-piperidinone,n-acetyl4-piperidone,pubchem19615,1-acetylpiperid-4-one
122563
1-acetylpiperidin-4-one
CC(=O)N1CCC(=O)CC1
Boiling Point 152°C
Color Yellow
Quantity 25g
Formula Weight 141.17
Physical Form Clear Liquid at 20°C
Percent Purity ≥98.0% (GC)
Chemical Name or Material 1-Acetyl-4-piperidone