Alfa Aesar™ 1-Acetylferrocene, 99+%

Manufacturer: Alfa Aesar™ 2295414

Quantity 25g

 View more versions of this product

Catalog No. AA2295414

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 1271-55-2
    Formula Weight 228.07
    MDL Number MFCD00001432
    UN Number UN3467
    Synonym Acetylferrocene; [(Acetylcyclopentadienyl)cyclopentadienyliron]
    Chemical Name or Material 1-Acetylferrocene

    Specifications

    Formula Weight 228.07
    Health Hazard 3 GHS P Statement
    P280-P301+P310-P361-P302+P350-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    IF ON SKIN: Gently wash with plenty of soap and water.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 1271-55-2
    Physical Form Crystalline
    MDL Number MFCD00001432
    UN Number UN3467
    Synonym Acetylferrocene; [(Acetylcyclopentadienyl)cyclopentadienyliron]
    Health Hazard 1 Danger
    Quantity 25g
    TSCA TSCA
    Melting Point 81° to 83°C
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC COMPOUND, SOLID, TOXIC, N.O.S.
    Health Hazard 2 GHS H Statement
    H300-H310
    Fatal if swallowed.
    Fatal in contact with skin.
    Chemical Name or Material 1-Acetylferrocene
    Assay ≥99%

    GHS H Statement
    H300-H310
    Fatal if swallowed.
    Fatal in contact with skin.

    GHS P Statement
    P280-P301+P310-P361-P302+P350-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    IF ON SKIN: Gently wash with plenty of soap and water.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit