Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Acetylindole-3-carboxaldehyde, 98%

Click to view available options
Quantity:
1g
5g
25g
22948-94-3
C11H9NO2
187.198
MFCD00039691
LCJLFGSKHBDOAY-UHFFFAOYSA-N
1-acetyl-1h-indole-3-carbaldehyde,n-acetylindole-3-carboxaldehyde,1-acetylindole-3-carboxaldehyde,1-acetyl-3-indolecarboxaldehyde,1h-indole-3-carboxaldehyde, 1-acetyl,1-acetyl-3-formylindole,zlchem 389,pubchem7227,n-acetylindole-3-aldehyde,acmc-1crl1
89915
1-acetylindole-3-carbaldehyde
CC(=O)N1C=C(C2=CC=CC=C21)C=O
22948-94-3
C11H9NO2
187.198
MFCD00039691
LCJLFGSKHBDOAY-UHFFFAOYSA-N
1-acetyl-1h-indole-3-carbaldehyde,n-acetylindole-3-carboxaldehyde,1-acetylindole-3-carboxaldehyde,1-acetyl-3-indolecarboxaldehyde,1h-indole-3-carboxaldehyde, 1-acetyl,1-acetyl-3-formylindole,zlchem 389,pubchem7227,n-acetylindole-3-aldehyde,acmc-1crl1
89915
1-acetylindole-3-carbaldehyde
CC(=O)N1C=C(C2=CC=CC=C21)C=O
Quantity 5g
Melting Point 160°C to 164°C
Formula Weight 187.2
Percent Purity 98%
Chemical Name or Material 1-Acetylindole-3-carboxaldehyde

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning