Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Acetylpiperazine, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
13889-98-0
C6H12N2O
128.17
MFCD00058676
PKDPUENCROCRCH-UHFFFAOYSA-N
1-acetylpiperazine,n-acetylpiperazine,1-piperazin-1-yl ethanone,1-piperazin-1-yl ethan-1-one,n-acetyl piperazine,acetylpiperazine,1-acetyl piperazine,piperazine, 1-acetyl,1-piperazin-1-yl-ethanone,1-piperazin-1-yl ethanon
83795
1-piperazin-1-ylethanone
CC(=O)N1CCNCC1
13889-98-0
C6H12N2O
128.17
MFCD00058676
PKDPUENCROCRCH-UHFFFAOYSA-N
1-acetylpiperazine,n-acetylpiperazine,1-piperazin-1-yl ethanone,1-piperazin-1-yl ethan-1-one,n-acetyl piperazine,acetylpiperazine,1-acetyl piperazine,piperazine, 1-acetyl,1-piperazin-1-yl-ethanone,1-piperazin-1-yl ethanon
83795
1-piperazin-1-ylethanone
CC(=O)N1CCNCC1
Melting Point 31°C to 34°C
Flash Point >110°C
Quantity 100g
Infrared Spectrum Authentic
Packaging Glass bottle
Color Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Beilstein 23,IV,201
Formula Weight 128.17
Physical Form Liquid After Melting
Percent Purity 98%
Chemical Name or Material 1-Acetylpiperazine
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 237-659-