Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Adamantanecarboxylic acid chloride, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
2094-72-6
C11H15ClO
198.69
MFCD00074724
MIBQYWIOHFTKHD-UHFFFAOYSA-N
1-adamantanecarbonyl chloride,adamantanecarbonyl chloride,1-adamantanecarbonylchloride,1-adamantoyl chloride,adamantoyl chloride,1-adamantanoic acid chloride,1-adamantanecarboxylic acid chloride,adamantane-1-carboxylic acid chloride,1-adamantylcarbonyl chloride,1-adamantyl carbonyl chloride
98915
adamantane-1-carbonyl chloride
C1C2CC3CC1CC(C2)(C3)C(=O)Cl
2094-72-6
C11H15ClO
198.69
MFCD00074724
MIBQYWIOHFTKHD-UHFFFAOYSA-N
1-adamantanecarbonyl chloride,adamantanecarbonyl chloride,1-adamantanecarbonylchloride,1-adamantoyl chloride,adamantoyl chloride,1-adamantanoic acid chloride,1-adamantanecarboxylic acid chloride,adamantane-1-carboxylic acid chloride,1-adamantylcarbonyl chloride,1-adamantyl carbonyl chloride
98915
adamantane-1-carbonyl chloride
C1C2CC3CC1CC(C2)(C3)C(=O)Cl
Quantity 5g
Melting Point 48.0°C to 52.0°C
Color White
Boiling Point 135.0°C to 136.0°C (10.0mmHg)
Packaging Glass bottle
Infrared Spectrum Authentic
Flash Point >112°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Fieser 02,15
Solubility Information Solubility in water: may decompose
Formula Weight 198.69
Physical Form Crystalline Solid
Percent Purity 97%
Chemical Name or Material 1-Adamantanecarboxylic acid chloride, 97%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 218-252-7

TSCA : TSCA