Alfa Aesar™ 1-Adamantanemethanol, 98%

Manufacturer: Alfa Aesar™ B2383103

Quantity 1g

 View more versions of this product

Catalog No. AAB2383103

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 770-71-8
    Molecular Formula C11H18O
    Formula Weight 166.26
    MDL Number MFCD00074751
    Beilstein 1853339
    Synonym 1-(Hydroxymethyl)adamantane
    Chemical Name or Material 1-Adamantanemethanol

    Specifications

    Formula Weight 166.26
    MDL Number MFCD00074751
    Melting Point 115° to 118°C
    Quantity 1g
    Molecular Formula C11H18O
    CAS 770-71-8
    Beilstein 1853339
    Synonym 1-(Hydroxymethyl)adamantane
    Chemical Name or Material 1-Adamantanemethanol
    Assay 98%

    Description

    Applications
    1-Adamantanemethanol was used in the synthesis of axially disubstituted silicon(IV) phthalocyanines. It is used as a reagent in the synthesis of adamantane and noradamantane based histone deacetylase (HDAC) inhibitors for the treatment of cancer. It is also used as a reagent in the synthesis of (1-adamantyl)methyl glycidyl ether, a versatile building block for living polymerization.

    Solubility
    Insoluble in water. Solubility in methanol (almost transparency).

    Notes
    Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit