Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Adamantanemethanol, 99%, ACROS Organics™

Click to view available options
Quantity:
2.5g
10g
50g
Packaging:
Glass bottle
770-71-8
C11H18O
166.26
MFCD00074751
MDVGOOIANLZFCP-UHFFFAOYSA-N
1-adamantanemethanol,adamantan-1-ylmethanol,1-adamantane methanol,1-hydroxymethyl adamantane,adamantane-1-methanol,adamantan-1-yl-methanol,1-hydroxymethyladamantane,tricyclo 3.3.1.1'3,7 dec-1-ylmethanol,adamantan-1-yl methanol,tricyclo 3.3.1.13,7 decanemethanol
64556
1-adamantylmethanol
C1C2CC3CC1CC(C2)(C3)CO
770-71-8
C11H18O
166.26
MFCD00074751
MDVGOOIANLZFCP-UHFFFAOYSA-N
1-adamantanemethanol,adamantan-1-ylmethanol,1-adamantane methanol,1-hydroxymethyl adamantane,adamantane-1-methanol,adamantan-1-yl-methanol,1-hydroxymethyladamantane,tricyclo 3.3.1.1'3,7 dec-1-ylmethanol,adamantan-1-yl methanol,tricyclo 3.3.1.13,7 decanemethanol
64556
1-adamantylmethanol
C1C2CC3CC1CC(C2)(C3)CO
Quantity 2.5g
Packaging Glass bottle
Infrared Spectrum Authentic
Melting Point 115.0°C to 118.0°C
Color White
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Solubility Information Solubility in water: .. Other solubilities: soluble in hexane and ethyl alcohol
Formula Weight 166.26
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material 1-Adamantanemethanol, 99%
Show More Show Less

EINECSNumber : 212-225-3

TSCA : TSCA