Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Adamantanemethylamine, 97%

Click to view available options
Quantity:
250mg
1g
5g
17768-41-1
C11H19N
165.28
MFCD00074750
XSOHXMFFSKTSIT-UHFFFAOYSA-N
1-adamantanemethylamine,adamantan-1-ylmethanamine,1-aminomethyl adamantane,1-aminomethyladamantane,1-aminomethyl-adamantane,1-1-adamantyl methanamine,1-adamantan-1-ylmethanamine,c-adamantan-1-yl-methylamine,1-adamantanemethyl amine,tricyclo 3.3.1.13,7 dec-1-ylmethylamine
86625
1-adamantylmethanamine
C1C2CC3CC1CC(C2)(C3)CN
17768-41-1
C11H19N
165.28
MFCD00074750
XSOHXMFFSKTSIT-UHFFFAOYSA-N
1-adamantanemethylamine,adamantan-1-ylmethanamine,1-aminomethyl adamantane,1-aminomethyladamantane,1-aminomethyl-adamantane,1-1-adamantyl methanamine,1-adamantan-1-ylmethanamine,c-adamantan-1-yl-methylamine,1-adamantanemethyl amine,tricyclo 3.3.1.13,7 dec-1-ylmethylamine
86625
1-adamantylmethanamine
C1C2CC3CC1CC(C2)(C3)CN
Sensitivity Air Sensitive
Refractive Index 1.5120
Quantity 1g
Density 0.933g/mL
Color Colorless to Yellow
Boiling Point 83°C to 85°C (0.3mmHg)
UN Number UN2735
Formula Weight 168.28
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 1-Adamantanemethylamine
Show More Show Less

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.