Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Adamantanemethylamine, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
17768-41-1
C11H19N
165.28
MFCD00074750
XSOHXMFFSKTSIT-UHFFFAOYSA-N
1-adamantanemethylamine,adamantan-1-ylmethanamine,1-aminomethyl adamantane,1-aminomethyladamantane,1-aminomethyl-adamantane,1-1-adamantyl methanamine,1-adamantan-1-ylmethanamine,c-adamantan-1-yl-methylamine,1-adamantanemethyl amine,tricyclo 3.3.1.13,7 dec-1-ylmethylamine
86625
1-adamantylmethanamine
C1C2CC3CC1CC(C2)(C3)CN
17768-41-1
C11H19N
165.28
MFCD00074750
XSOHXMFFSKTSIT-UHFFFAOYSA-N
1-adamantanemethylamine,adamantan-1-ylmethanamine,1-aminomethyl adamantane,1-aminomethyladamantane,1-aminomethyl-adamantane,1-1-adamantyl methanamine,1-adamantan-1-ylmethanamine,c-adamantan-1-yl-methylamine,1-adamantanemethyl amine,tricyclo 3.3.1.13,7 dec-1-ylmethylamine
86625
1-adamantylmethanamine
C1C2CC3CC1CC(C2)(C3)CN
Specific Gravity 0.93
Boiling Point 83.0°C to 85.0°C (0.3mmHg)
Quantity 1g
Packaging Glass bottle
Flash Point 92°C
Refractive Index 1.5127 to 1.5147
Infrared Spectrum Authentic
Color Colorless to Yellow
Density 0.9300g/mL
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 165.28
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 1-Adamantanemethylamine
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 241-752-1