Alfa Aesar™ 1-Adamantyl methyl ketone, 99%

Manufacturer: Alfa Aesar™ H5588914

Quantity 25g

 View more versions of this product

Catalog No. AAH5588914

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1660-04-4
    Molecular Formula C12H18O
    Formula Weight 178.27
    MDL Number MFCD00074739
    Chemical Name or Material 1-Adamantyl methyl ketone

    Specifications

    Formula Weight 178.27
    Molecular Formula C12H18O
    MDL Number MFCD00074739
    Quantity 25g
    Flash Point 108°C (226°F)
    CAS 1660-04-4
    Melting Point 53° to 55°C
    Chemical Name or Material 1-Adamantyl methyl ketone
    Assay 99%

    Description

    Applications
    1-Adamantyl methyl ketone, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

    Solubility
    Soluble in methanol- very faint turbidity. Insoluble in water.

    Notes
    Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents. Stable under normal temperatures and pressures.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit