Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Allylpiperazine, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
13961-36-9
C7H14N2
126.2
MFCD00167970
ZWAQJGHGPPDZSF-UHFFFAOYSA-N
1-allylpiperazine,1-allyl-piperazine,1-allyl piperazine,1-prop-2-en-1-yl piperazine,n-allyl piperazine,piperazine,1-2-propen-1-yl,prop-2-enylpiperazine,allylpiperazin,allylpiperazine,1-allylpiperazin
806422
1-prop-2-enylpiperazine
C=CCN1CCNCC1
13961-36-9
C7H14N2
126.2
MFCD00167970
ZWAQJGHGPPDZSF-UHFFFAOYSA-N
1-allylpiperazine,1-allyl-piperazine,1-allyl piperazine,1-prop-2-en-1-yl piperazine,n-allyl piperazine,piperazine,1-2-propen-1-yl,prop-2-enylpiperazine,allylpiperazin,allylpiperazine,1-allylpiperazin
806422
1-prop-2-enylpiperazine
C=CCN1CCNCC1
Specific Gravity 1
Refractive Index 1.4830 to 1.4850 (20°C, 589nm)
Quantity 5g
Flash Point 51.7°C
Density 1.0000g/mL
Packaging Glass bottle
Color Yellow
Boiling Point 181.0°C to 182.0°C
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Formula Weight 126.2
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 1-Allylpiperazine
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger