Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Amino-1-cyclohexanecarboxylic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
2756-85-6
C7H13NO2
143.19
MFCD00001487
WOXWUZCRWJWTRT-UHFFFAOYSA-N
1-aminocyclohexanecarboxylic acid,homocycloleucine,1-amino-1-cyclohexanecarboxylic acid,cyclohexanecarboxylic acid, 1-amino,alpha-aminocyclohexanecarboxylic acid,alpha-aminocyclohexanecarboxylic aicd,unii-1se3hd391h,1-amino-1-cyclohexane carboxylic acid,.alpha.-aminocyclohexanecarboxylic acid,.alpha.-aminocyclohexanecarboxylic aicd
1366
CHEBI:86534
1-aminocyclohexane-1-carboxylic acid
C1CCC(CC1)(C(=O)O)N
2756-85-6
C7H13NO2
143.19
MFCD00001487
WOXWUZCRWJWTRT-UHFFFAOYSA-N
1-aminocyclohexanecarboxylic acid,homocycloleucine,1-amino-1-cyclohexanecarboxylic acid,cyclohexanecarboxylic acid, 1-amino,alpha-aminocyclohexanecarboxylic acid,alpha-aminocyclohexanecarboxylic aicd,unii-1se3hd391h,1-amino-1-cyclohexane carboxylic acid,.alpha.-aminocyclohexanecarboxylic acid,.alpha.-aminocyclohexanecarboxylic aicd
1366
CHEBI:86534
1-aminocyclohexane-1-carboxylic acid
C1CCC(CC1)(C(=O)O)N
Melting Point >300°C
Infrared Spectrum Authentic
Packaging Glass bottle
Color White
Quantity 100g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Beilstein 14, 299
Formula Weight 143.19
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 1-Amino-1-cyclohexanecarboxylic acid, 98%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 220-411-0

RTECSNumber : GU8393000

TSCA : TSCA