Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1-Amino-1-methylpropyl)phosphonic acid, hydrate, 97%, Thermo Scientific™

Catalog Number AC344870010
Click to view available options
Quantity:
1 g
Packaging:
Glass Bottle
79014-65-6
C4H14NO4P
171.133
MFCD03791053
XOFGFZIYHLOEQZ-UHFFFAOYSA-N
2-aminobutan-2-yl phosphonic acid hydrate, 2-aminobutan-2-ylphosphonic acid hydrate, phosphonic acid, 1-amino-1-methylpropyl-9ci, 2-aminobutan-2-yl phosphonic acid-water 1/1, 1-amino-1-methylpropyl phosphonic acid hydrate
53393191
2-aminobutan-2-ylphosphonic acid;hydrate
CCC(C)(N)P(=O)(O)O.O
79014-65-6
C4H14NO4P
171.133
MFCD03791053
XOFGFZIYHLOEQZ-UHFFFAOYSA-N
2-aminobutan-2-yl phosphonic acid hydrate, 2-aminobutan-2-ylphosphonic acid hydrate, phosphonic acid, 1-amino-1-methylpropyl-9ci, 2-aminobutan-2-yl phosphonic acid-water 1/1, 1-amino-1-methylpropyl phosphonic acid hydrate
53393191
2-aminobutan-2-ylphosphonic acid;hydrate
CCC(C)(N)P(=O)(O)O.O

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Linear Formula CH3CH2C(CH3)(NH2)P(O)(OH)2·xH2O
Quantity 1 g
Formula Weight 153.12
Percent Purity ≥96.0% (on dry substance)
Physical Form Powder
Chemical Name or Material (1-Amino-1-methylpropyl)phosphonic acid, hydrate, 97%
Show More Show Less
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit