Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1-Amino-1-phenylmethyl)phosphonic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
18108-22-0
C7H10NO3P
MFCD00014809
α-Aminobenzylphosphonic acid
18108-22-0
C7H10NO3P
MFCD00014809
α-Aminobenzylphosphonic acid
Quantity 1g
Melting Point 279°C to 284°C
Packaging Glass bottle
Color White
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula C6H5CH(NH)P(O)(OH)
Formula Weight 187.13
Physical Form Powder or Cyrstals
Percent Purity ≥96.0%
Chemical Name or Material (1-Amino-1-phenylmethyl)phosphonic acid, 97%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.