Alfa Aesar™ 1-Amino-2-naphthol hydrochloride, 97%

Manufacturer: Alfa Aesar™ B2449214

Quantity 25g

 View more versions of this product

Catalog No. AAB2449214

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1198-27-2
    Molecular Formula C10H9NO·HCl
    Formula Weight 195.65
    MDL Number MFCD00012561
    Beilstein 3699817
    Synonym 2-Hydroxy-1-naphthylamine hydrochloride; 2-Hydroxy-1-naphthylamine hydrochloride
    Chemical Name or Material 1-Amino-2-naphthol hydrochloride

    Specifications

    Formula Weight 195.65
    TSCA TSCA
    Melting Point ≈276°C (dec.)
    Quantity 25g
    Molecular Formula C10H9NO·HCl
    MDL Number MFCD00012561
    Beilstein 3699817
    Synonym 2-Hydroxy-1-naphthylamine hydrochloride; 2-Hydroxy-1-naphthylamine hydrochloride
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H341-H315-H319-H335
    Suspected of causing genetic defects.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    RTECS Number QL3335000
    CAS 1198-27-2
    Health Hazard 3 GHS P Statement
    P261-P280-P281-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 1-Amino-2-naphthol hydrochloride
    Assay 97%

    GHS H Statement
    H341-H315-H319-H335
    Suspected of causing genetic defects.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P281-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit