Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Amino-2-naphthol hydrochloride, 97%

Click to view available options
Quantity:
1g
5g
25g
1198-27-2
C10H10ClNO
195.646
MFCD00012561
DEKREBCFNLUULC-UHFFFAOYSA-N
1-amino-2-naphthol hydrochloride,1-aminonaphthalen-2-ol hydrochloride,1-amino-2-naphthol hcl,unii-xm2lzu54ow,2-naphthalenol, 1-amino-, hydrochloride 1:1,2-naphthalenol, 1-amino-, hydrochloride,xm2lzu54ow,1-aminonaphthalen-2-ol, chloride,timtec-bb sbb000347,acmc-209a4b
518469
1-aminonaphthalen-2-ol;hydrochloride
C1=CC=C2C(=C1)C=CC(=C2N)O.Cl
1198-27-2
C10H10ClNO
195.646
MFCD00012561
DEKREBCFNLUULC-UHFFFAOYSA-N
1-amino-2-naphthol hydrochloride,1-aminonaphthalen-2-ol hydrochloride,1-amino-2-naphthol hcl,unii-xm2lzu54ow,2-naphthalenol, 1-amino-, hydrochloride 1:1,2-naphthalenol, 1-amino-, hydrochloride,xm2lzu54ow,1-aminonaphthalen-2-ol, chloride,timtec-bb sbb000347,acmc-209a4b
518469
1-aminonaphthalen-2-ol;hydrochloride
C1=CC=C2C(=C1)C=CC(=C2N)O.Cl
Melting Point ~276°C (decomposition)
Sensitivity Light sensitive
Quantity 5g
Beilstein 3699817
Formula Weight 195.65
Percent Purity 97%
Chemical Name or Material 1-Amino-2-naphthol hydrochloride

GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c

H315-H319-H335-H341

EINECSNumber : 214-830-8

RTECSNumber : QL3335000

TSCA : Yes

Recommended Storage : Ambient temperatures