Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Amino-2-naphthol Hydrochloride 97.0+%, TCI America™

Click to view available options
Quantity:
25g
1198-27-2
C10H10ClNO
195.646
MFCD00012561
DEKREBCFNLUULC-UHFFFAOYSA-N
1-amino-2-naphthol hydrochloride,1-aminonaphthalen-2-ol hydrochloride,1-amino-2-naphthol hcl,unii-xm2lzu54ow,2-naphthalenol, 1-amino-, hydrochloride 1:1,2-naphthalenol, 1-amino-, hydrochloride,xm2lzu54ow,1-aminonaphthalen-2-ol, chloride,timtec-bb sbb000347,acmc-209a4b
518469
1-aminonaphthalen-2-ol;hydrochloride
C1=CC=C2C(=C1)C=CC(=C2N)O.Cl
1198-27-2
C10H10ClNO
195.646
MFCD00012561
DEKREBCFNLUULC-UHFFFAOYSA-N
1-amino-2-naphthol hydrochloride,1-aminonaphthalen-2-ol hydrochloride,1-amino-2-naphthol hcl,unii-xm2lzu54ow,2-naphthalenol, 1-amino-, hydrochloride 1:1,2-naphthalenol, 1-amino-, hydrochloride,xm2lzu54ow,1-aminonaphthalen-2-ol, chloride,timtec-bb sbb000347,acmc-209a4b
518469
1-aminonaphthalen-2-ol;hydrochloride
C1=CC=C2C(=C1)C=CC(=C2N)O.Cl
Color Purple
Quantity 25g
Formula Weight 195.65
Physical Form Crystal-Powder at 20°C
Percent Purity ≥97.0% (HPLC,N)
Chemical Name or Material 1-Amino-2-naphthol Hydrochloride

EINECSNumber : (4)-0363

RTECSNumber : QL3335000

TSCA : Yes