Alfa Aesar™ 1-Amino-4-(2-hydroxyethyl)piperazine, 96%

Manufacturer: Alfa Aesar™ H55134MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH55134MD

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 3973-70-4
    Formula Weight 145.2
    MDL Number MFCD00006155
    Synonym 4-Amino-1-piperazineethanol
    Chemical Name or Material 1-Amino-4-(2-hydroxyethyl)piperazine

    Specifications

    Melting Point 105° to 107°C
    Formula Weight 145.2
    Physical Form Powder
    MDL Number MFCD00006155
    Color White to pale brown
    Quantity 250mg
    CAS 3973-70-4
    Synonym 4-Amino-1-piperazineethanol
    Chemical Name or Material 1-Amino-4-(2-hydroxyethyl)piperazine
    Assay 96%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit