Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Amino-4-hydroxyanthraquinone, 96%

Click to view available options
Quantity:
5g
25g
100g
116-85-8
C14H9NO3
239.23
MFCD00001223
AQXYVFBSOOBBQV-UHFFFAOYSA-N
1-amino-4-hydroxyanthraquinone,disperse red 15,calcosyn pink b,celutate pink b,nacelan pink b,amacel pink b,celutate pink bn,celutate pink by,cibacete red 3b,disperse red 2s
8323
1-amino-4-hydroxyanthracene-9,10-dione
C1=CC=C2C(=C1)C(=O)C3=C(C=CC(=C3C2=O)O)N
116-85-8
C14H9NO3
239.23
MFCD00001223
AQXYVFBSOOBBQV-UHFFFAOYSA-N
1-amino-4-hydroxyanthraquinone,disperse red 15,calcosyn pink b,celutate pink b,nacelan pink b,amacel pink b,celutate pink bn,celutate pink by,cibacete red 3b,disperse red 2s
8323
1-amino-4-hydroxyanthracene-9,10-dione
C1=CC=C2C(=C1)C(=O)C3=C(C=CC(=C3C2=O)O)N
Applications
The use of the inclusion complex of 1-amino-4-hydroxyanthraquinone (AHA) in the internal cavity of β-cyclodextrin confers increased sensitivity, selectivity and detection limit, and allows the determination of 10-70 ng ml-1 beryllium, compared to 60-500 ng ml-1 in the absence of the cyclodextrin.

Solubility
Soluble in water, ethanol, acetone, benzene.

Notes
Store at room temperature. Incompatible with oxidizing agents.
Quantity 100g
Melting Point 207°C to 209°C
Formula Weight 239.23
Percent Purity 96%
Chemical Name or Material 1-Amino-4-hydroxyanthraquinone

GHS H Statement
H341
Suspected of causing genetic defects.

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA