Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Aminobenzotriazole, 98%, ACROS Organics™

Click to view available options
Quantity:
100mg
250mg
Packaging:
Glass bottle
1614-12-6
C6H6N4
134.14
MFCD00132902
JCXKHYLLVKZPKE-UHFFFAOYSA-N
1-aminobenzotriazole,1h-benzotriazol-1-amine,1h-benzo d 1,2,3 triazol-1-amine,1-benzotriazolamine,1-abtz,1h-1,2,3-benzotriazol-1-amine,1h-1,2,3-benzotriazol-1-ylamine,1,2,3-benzotriazol-1-amine,benzotriazolylamine,1-benzotriazolylamine
1367
benzotriazol-1-amine
C1=CC=C2C(=C1)N=NN2N
1614-12-6
C6H6N4
134.14
MFCD00132902
JCXKHYLLVKZPKE-UHFFFAOYSA-N
1-aminobenzotriazole,1h-benzotriazol-1-amine,1h-benzo d 1,2,3 triazol-1-amine,1-benzotriazolamine,1-abtz,1h-1,2,3-benzotriazol-1-amine,1h-1,2,3-benzotriazol-1-ylamine,1,2,3-benzotriazol-1-amine,benzotriazolylamine,1-benzotriazolylamine
1367
benzotriazol-1-amine
C1=CC=C2C(=C1)N=NN2N
Color Beige to Yellow or Brown
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 81°C to 85°C
Quantity 250mg
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in chloroform,carbon tetrachloride,,soluble in actic acid
Formula Weight 134.14
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 1-Aminobenzotriazole, 98%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

TSCA : TSCA