Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Aminocyclopropanecarboxylic acid hydrochloride, 97%, ACROS Organics™

Click to view available options
Quantity:
100mg
500mg
Packaging:
Glass bottle
68781-13-5
C4H7NO2·ClH
137.57
MFCD00012545
WTZRHZRMLYNBRM-UHFFFAOYSA-N
1-aminocyclopropanecarboxylic acid hydrochloride,1-aminocyclopropane-1-carboxylic acid hydrochloride,1-amino-cyclopropane-1-carboxylic acid hydrochloride,1-aminocyclopropanecarboxylic acid hcl,cyclopropanecarboxylic acid, 1-amino-, hydrochloride,1-aminocyclopropanecarboxylic acid, chloride,acpc-hcl,pubchem13872,ksc352q0j,1-amino-cyclopropane-1-carboxylic acid, hcl
11957445
1-aminocyclopropane-1-carboxylic acid;hydrochloride
C1CC1(C(=O)O)N.Cl
68781-13-5
C4H7NO2·ClH
137.57
MFCD00012545
WTZRHZRMLYNBRM-UHFFFAOYSA-N
1-aminocyclopropanecarboxylic acid hydrochloride,1-aminocyclopropane-1-carboxylic acid hydrochloride,1-amino-cyclopropane-1-carboxylic acid hydrochloride,1-aminocyclopropanecarboxylic acid hcl,cyclopropanecarboxylic acid, 1-amino-, hydrochloride,1-aminocyclopropanecarboxylic acid, chloride,acpc-hcl,pubchem13872,ksc352q0j,1-amino-cyclopropane-1-carboxylic acid, hcl
11957445
1-aminocyclopropane-1-carboxylic acid;hydrochloride
C1CC1(C(=O)O)N.Cl
Melting Point 221.6°C to 225.0°C
Quantity 500mg
Color White
Infrared Spectrum Authentic
Packaging Glass bottle
Elemental Analysis 33.52 to 36.31% C, 5.63 to 6.09% H, 9.77 to 10.59% N
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 137.57
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 1-Aminocyclopropanecarboxylic acid hydrochloride
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning