Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1-Aminoethyl)phosphonic Acid, TCI America™

Click to view available options
Quantity:
1g
6323-97-3
C2H8NO3P
125.064
MFCD00008068
UIQSKEDQPSEGAU-UHFFFAOYSA-N
1-aminoethyl phosphonic acid,1-ammonium-ethanephosphonic acid anion,phosphonic acid, 1-aminoethyl,1-phosphonoethylamine,acmc-20aphh,acmc-209ne9,uiqskedqpsegau-uhfffaoysa,1-amino-ethyl-phosphonic acid,dl-1-aminoethyl phosphonic acid
97709
1-aminoethylphosphonic acid
CC(N)P(=O)(O)O
6323-97-3
C2H8NO3P
125.064
MFCD00008068
UIQSKEDQPSEGAU-UHFFFAOYSA-N
1-aminoethyl phosphonic acid,1-ammonium-ethanephosphonic acid anion,phosphonic acid, 1-aminoethyl,1-phosphonoethylamine,acmc-20aphh,acmc-209ne9,uiqskedqpsegau-uhfffaoysa,1-amino-ethyl-phosphonic acid,dl-1-aminoethyl phosphonic acid
97709
1-aminoethylphosphonic acid
CC(N)P(=O)(O)O
Quantity 1g
Color White
Formula Weight 125.06
Physical Form Crystal-Powder at 20°C
Percent Purity ≥95.0% (T)
Chemical Name or Material (1-Aminoethyl)phosphonic Acid

EINECSNumber : (9)-0067

TSCA : No